CAS RN 1678513-97-7
CAS RN 1678513-97-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BMeS-p-A 95.0+%, TCI America™
CAS: 1678513-97-7 Molecular Formula: C8H12N2O4S2 Synonym: 1,4-Diamino-2,5-bis(methylsulfonyl)benzene, 2,5-Bis(methylsulfonyl)-1,4-phenylenediamine