accessibility menu, dialog, popup

UserName

CH3OHOOCH3

CAS RN 16777-87-0

IUPAC Name: 3,3-diethoxypropan-1-ol
Synonyms: 3,3-diethoxypropan-1-ol
Molecular Weight (g/mol): 148.2
Molecular Formula: C7H16O3
InChi Key: ASERXEZXVIJBRO-UHFFFAOYSA-N
SMILES: CCOC(CCO)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)