accessibility menu, dialog, popup

UserName

CAS RN 16743-33-2

OOOOZnH2OH2OCF3CF3CF3CF3

CAS RN 16743-33-2

IUPAC Name: zinc(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) dihydrate
Synonyms: zinc(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate) dihydrate
Molecular Weight (g/mol): 515.51
Molecular Formula: C10H6F12O6Zn
InChi Key: HHDNHEKLDWSJAJ-LJDKTGGESA-L
SMILES: O.O.[Zn++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results