CAS RN 16713-66-9
IUPAC Name:
2-[1-(carboxymethyl)cyclopentyl]acetic acid
Synonyms:
[1-(carboxymethyl)cyclopentyl]acetic acid
Molecular Weight (g/mol):
186.21
Molecular Formula:
C9H14O4
InChi Key:
FWPVKDFOUXHOKQ-UHFFFAOYSA-N
SMILES:
OC(=O)CC1(CC(O)=O)CCCC1
1
–
1
of
1
results
1
1,1-Cyclopentanediacetic Acid 98.0+%, TCI America™
CAS: 16713-66-9 Molecular Formula: C9H14O4 Molecular Weight (g/mol): 186.21 MDL Number: MFCD00001388 InChI Key: FWPVKDFOUXHOKQ-UHFFFAOYSA-N Synonym: 1,1-cyclopentanediacetic acid,3,3-tetramethyleneglutaric acid,cyclopentane-1,1-diacetic acid,cyclopentanediacetic acid,tetramethyleneglutaric acid,tetramethylene glutaric acid,2-1-carboxymethyl cyclopentyl acetic acid,1,1-cyclopentanediaceticacid,chembl80679,3,3'-tetramethylene glutaric acid PubChem CID: 85558 IUPAC Name: 2-[1-(carboxymethyl)cyclopentyl]acetic acid SMILES: OC(=O)CC1(CC(O)=O)CCCC1