accessibility menu, dialog, popup

UserName

CAS RN 16687-60-8

IUPAC Name: 5-(4-nitrophenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(4-nitrophenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 191.15
Molecular Formula: C7H5N5O2
InChi Key: MIUOBAHGBPSRKY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)