CAS RN 16606-55-6
CAS RN 16606-55-6
IUPAC Name:
4-methyl-1,3-dioxolan-2-one
Synonyms:
propylene carbonate arconate 5000 propylene carbonate preparation 4-methyldioxalone-2 propylene glycol cyclic carbonate propylenester kyseliny uhlicite carbonic acid, cyclic propylene ester carbonic acid, cyclic propylene ether 1,2-propylene carbonate dipropylene carbonate
Molecular Weight (g/mol):
102.09
Molecular Formula:
C4H6O3
InChi Key:
RUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILES:
CC1COC(=O)O1
1
–
1
of
1
results
1
(R)-Propylene Carbonate 98.0+%, TCI America™
CAS: 16606-55-6 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.089 MDL Number: MFCD00798265 InChI Key: RUOJZAUFBMNUDX-GSVOUGTGSA-N Synonym: (R)-4-Methyl-1,3-dioxolan-2-one PubChem CID: 641813 IUPAC Name: (4R)-4-methyl-1,3-dioxolan-2-one SMILES: CC1COC(=O)O1