accessibility menu, dialog, popup

UserName

H3CCH3(S)NH3OOCH3CH3H3C

CAS RN 166023-30-9

IUPAC Name: (3S)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Synonyms: (3S)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Molecular Weight (g/mol): 202.32
Molecular Formula: C11H24NO2
InChi Key: WBOPVTYXLFEVGP-VIFPVBQESA-O
SMILES: CC(C)C[C@H]([NH3+])CC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results