accessibility menu, dialog, popup

UserName

CAS RN 1660137-09-6

NbOOOOOOOHOOHOOOOOHOOHOOOHO

CAS RN 1660137-09-6

IUPAC Name: tetrakis[(carboxycarbonyl)oxy]niobio hydrogen oxalate
Synonyms: tetrakis[(carboxycarbonyl)oxy]niobio hydrogen oxalate
Molecular Weight (g/mol): 538.04
Molecular Formula: C10H5NbO20
InChi Key: AXIFGFAGYFPNFC-UHFFFAOYSA-I
SMILES: OC(=O)C(=O)O[Nb](OC(=O)C(O)=O)(OC(=O)C(O)=O)(OC(=O)C(O)=O)OC(=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)