accessibility menu, dialog, popup

UserName

NOOON

CAS RN 1659-31-0

IUPAC Name: bis(pyridin-2-yl) carbonate
Synonyms: bis(pyridin-2-yl) carbonate
Molecular Weight (g/mol): 216.2
Molecular Formula: C11H8N2O3
InChi Key: GCSAXWHQFYOIFE-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=N1)OC1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results