accessibility menu, dialog, popup

UserName

CH3CH2N

CAS RN 16529-56-9

IUPAC Name: 2-methylbut-3-enenitrile
Synonyms: 3-butenenitrile, 2-methyl-
Molecular Weight (g/mol): 81.12
Molecular Formula: C5H7N
InChi Key: WBAXCOMEMKANRN-UHFFFAOYNA-N
SMILES: CC(C=C)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results