accessibility menu, dialog, popup

UserName

CAS RN 164724-35-0

NN

CAS RN 164724-35-0

IUPAC Name: N4,N4,N4',N4'-tetrakis({[1,1'-biphenyl]-4-yl})-[1,1'-biphenyl]-4,4'-diamine
Synonyms: N4,N4,N4',N4'-tetrakis({[1,1'-biphenyl]-4-yl})-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 793.03
Molecular Formula: C60H44N2
InChi Key: WXAIEIRYBSKHDP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results