accessibility menu, dialog, popup

UserName

CAS RN 164513-38-6

IUPAC Name: 5-bromo-4-methyl-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-4-methyl-1H-pyridin-2-one
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: BYGNXKDNIFHZHQ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results