CAS RN 1644-68-4
IUPAC Name:
2,3-bis(trifluoromethyl)pyridine
Synonyms:
2,3-bis(trifluoromethyl)pyridine
Molecular Weight (g/mol):
215.1
Molecular Formula:
C7H3F6N
InChi Key:
RRNXYHYDSDAOFW-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=C(N=CC=C1)C(F)(F)F
1
–
1
of
1
results
1
2,3-Bis(trifluoromethyl)pyridine 95.0+%, TCI America™
CAS: 1644-68-4 Molecular Formula: C7H3F6N Molecular Weight (g/mol): 215.098 MDL Number: MFCD03093763 InChI Key: RRNXYHYDSDAOFW-UHFFFAOYSA-N Synonym: 2,3-bis trifluoromethyl pyridine,pyridine, 2,3-bis trifluoromethyl,pubchem7694,acmc-209dra,bis trifluoromethyl pyridine,2,3-bis trifluoromethyl pyridine #,pyridine,2,3-bis trifluoromethyl PubChem CID: 589865 IUPAC Name: 2,3-bis(trifluoromethyl)pyridine SMILES: C1=CC(=C(N=C1)C(F)(F)F)C(F)(F)F