accessibility menu, dialog, popup

UserName

CAS RN 163927-32-0

IUPAC Name: 7-(3-isothiocyanatopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms: 7-(3-isothiocyanatopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol): 353.42
Molecular Formula: C13H15N5O3S2
InChi Key: YMXYIIXTKUQWPC-UHFFFAOYNA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(N2CCC(C2)N=C=S)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results