accessibility menu, dialog, popup

UserName

CH3

CAS RN 16327-42-7

IUPAC Name: 5-methyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Synonyms: 5-methyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular Weight (g/mol): 146.23
Molecular Formula: C11H14
InChi Key: VRLWYYZVFNNMET-UHFFFAOYNA-N
SMILES: CC1C=CC2C3CC(C=C3)C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results