CAS RN 16321-99-6
CAS RN 16321-99-6
IUPAC Name:
[3,4-bis(acetyloxy)-5-(2-amino-6-chloro-9H-purin-9-yl)oxolan-2-yl]methyl acetate
Synonyms:
[3,4-bis(acetyloxy)-5-(2-amino-6-chloropurin-9-yl)oxolan-2-yl]methyl acetate
Molecular Weight (g/mol):
427.8
Molecular Formula:
C16H18ClN5O7
InChi Key:
HLVGVJFVAKPFOZ-UHFFFAOYNA-N
SMILES:
CC(=O)OCC1OC(C(OC(C)=O)C1OC(C)=O)N1C=NC2=C(Cl)N=C(N)N=C12
1
–
1
of
1
results
1
2',3',5'-Tri-O-acetyl-2-amino-6-chloropurine Riboside 97.0+%, TCI America™
CAS: 16321-99-6 Molecular Formula: C16H18ClN5O7 Molecular Weight (g/mol): 427.80 MDL Number: MFCD03425473 InChI Key: HLVGVJFVAKPFOZ-UHFFFAOYNA-N Synonym: 2-Amino-6-chloro-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine PubChem CID: 10788628 IUPAC Name: [3,4-bis(acetyloxy)-5-(2-amino-6-chloro-9H-purin-9-yl)oxolan-2-yl]methyl acetate SMILES: CC(=O)OCC1OC(C(OC(C)=O)C1OC(C)=O)N1C=NC2=C(Cl)N=C(N)N=C12