CAS RN 1631-70-5
IUPAC Name:
benzyldimethylsilyl
Synonyms:
benzyldimethylsilyl
Molecular Weight (g/mol):
149.29
Molecular Formula:
C9H13Si
InChi Key:
SKDKWDVYZOKBPL-UHFFFAOYSA-N
SMILES:
C[Si](C)CC1=CC=CC=C1
1
–
1
of
1
results
1
Benzyldimethylsilane, 96%
CAS: 1631-70-5 Molecular Formula: C9H13Si Molecular Weight (g/mol): 149.288 MDL Number: MFCD00039792 InChI Key: SKDKWDVYZOKBPL-UHFFFAOYSA-N Synonym: benzyldimethylsilane,dimethyl phenylmethyl silane,silane, dimethyl phenylmethyl,benzyldimethylsilyl,benzyl dimethyl silicon,benzene, dimethylsilyl methyl PubChem CID: 6328719 IUPAC Name: benzyl(dimethyl)silicon SMILES: C[Si](C)CC1=CC=CC=C1