accessibility menu, dialog, popup

UserName

ClClSClOO

CAS RN 16271-33-3

IUPAC Name: 2,4-dichlorobenzene-1-sulfonyl chloride
Synonyms: 2,4-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 245.5
Molecular Formula: C6H3Cl3O2S
InChi Key: FDTPBIKNYWQLAE-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(Cl)=C1)S(Cl)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)