accessibility menu, dialog, popup

UserName

CAS RN 16205-44-0

H3COONN

CAS RN 16205-44-0

IUPAC Name: ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
Synonyms: ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: IDHMFSBXNNIWCT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C2C=CC=CN2N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)