CAS RN 1614-11-5
IUPAC Name:
2H-1,2,3-benzotriazol-2-amine
Synonyms:
1,2,3-benzotriazol-2-amine
Molecular Weight (g/mol):
134.14
Molecular Formula:
C6H6N4
InChi Key:
YYZVLEJDGSWXII-UHFFFAOYSA-N
SMILES:
NN1N=C2C=CC=CC2=N1
1
–
1
of
1
results
1
2-Aminobenzotriazole 98.0+%, TCI America™
CAS: 1614-11-5 Molecular Formula: C6H6N4 Molecular Weight (g/mol): 134.142 MDL Number: MFCD00724002 InChI Key: YYZVLEJDGSWXII-UHFFFAOYSA-N Synonym: 2-Benzotriazolamine PubChem CID: 295514 IUPAC Name: benzotriazol-2-amine SMILES: C1=CC2=NN(N=C2C=C1)N