CAS RN 1576-84-7
CAS RN 1576-84-7
IUPAC Name:
but-3-en-1-yl acetate
Synonyms:
but-3-en-1-yl acetate
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
IEKXSSZASGLISC-UHFFFAOYSA-N
SMILES:
CC(=O)OCCC=C
1
–
1
of
1
results
1
3-Butenyl Acetate 98.0+%, TCI America™
CAS: 1576-84-7 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00086596 InChI Key: IEKXSSZASGLISC-UHFFFAOYSA-N Synonym: 4-Acetoxy-1-butene, Acetic Acid 3-Butenyl Ester PubChem CID: 74095 IUPAC Name: but-3-enyl acetate SMILES: CC(=O)OCCC=C