accessibility menu, dialog, popup

UserName

BNNH3CH3CH3CNCH3CH3CH3FF

CAS RN 157410-23-6

IUPAC Name: 8-cyano-2,2-difluoro-4,5,6,10,11,12-hexamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Synonyms: 8-cyano-2,2-difluoro-4,5,6,10,11,12-hexamethyl-1λ⁵,3-diaza-2-boratricyclo[7.3.0.0³,⁷]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide
Molecular Weight (g/mol): 301.15
Molecular Formula: C16H18BF2N3
InChi Key: LXNVCQRKLGCOPW-UHFFFAOYSA-N
SMILES: CC1=C(C)C(C)=C2N1[B-](F)(F)[N+]1=C(C)C(C)=C(C)C1=C2C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results