accessibility menu, dialog, popup

UserName

H3CNBr

CAS RN 156972-74-6

IUPAC Name: 3-bromo-9-hexyl-9H-carbazole
Synonyms: 3-bromo-9-hexylcarbazole
Molecular Weight (g/mol): 330.27
Molecular Formula: C18H20BrN
InChi Key: HYUCKXZMLJFXSS-UHFFFAOYSA-N
SMILES: CCCCCCN1C2=C(C=CC=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results