accessibility menu, dialog, popup

UserName

CH3OH3C

CAS RN 1567-75-5

IUPAC Name: 1-(1-methylcyclopropyl)ethan-1-one
Synonyms: 1-(1-methylcyclopropyl)ethanone
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: OQBCJXUAQQMTRW-UHFFFAOYSA-N
SMILES: CC(=O)C1(C)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)