accessibility menu, dialog, popup

UserName

CH3SiCH3H3CSiCH3CH3Cl

CAS RN 1560-28-7

IUPAC Name: 1-chloro-1,1,2,2,2-pentamethyldisilane
Synonyms: 1-chloro-1,1,2,2,2-pentamethyldisilane
Molecular Weight (g/mol): 166.8
Molecular Formula: C5H15ClSi2
InChi Key: GJCAUTWJWBFMFU-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[Si](C)(C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results