accessibility menu, dialog, popup

UserName

CAS RN 156-62-7

CaH2NN

CAS RN 156-62-7

IUPAC Name: calcium aminoformonitrile
Synonyms: calcium cyanamide
Molecular Weight (g/mol): 82.12
Molecular Formula: CH2CaN2
InChi Key: STRTXDFFNXSZQB-UHFFFAOYSA-N
SMILES: [Ca++].NC#N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results