CAS RN 15523-24-7
IUPAC Name:
sodium tetraethylboranuide
Synonyms:
sodium tetraethylboranuide
Molecular Weight (g/mol):
150.05
Molecular Formula:
C8H20BNa
InChi Key:
SZSBMTRYJRHYNI-UHFFFAOYSA-N
SMILES:
[Na+].CC[B-](CC)(CC)CC
1
–
1
of
1
results
1
Sodium tetraethylborate, 97%, pure, Thermo Scientific Chemicals
CAS: 15523-24-7 Molecular Formula: C8H20BNa Molecular Weight (g/mol): 150.04 MDL Number: MFCD00061547 InChI Key: SZSBMTRYJRHYNI-UHFFFAOYSA-N Synonym: sodium tetraethylborate,sodium tetraethylboranuide,borate 1-,tetraethyl-, sodium 1:1,sodium tetraethylborate 1-,et4bna,sodiotetraethylboron v,borate 1-, tetraethyl-, sodium,sodium tetraethylborate 1g PubChem CID: 23681030 IUPAC Name: sodium;tetraethylboranuide SMILES: [B-](CC)(CC)(CC)CC.[Na+]