CAS RN 15484-80-7
CAS RN 15484-80-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Vinyl 4-tert-Butylbenzoate 98.0+%, TCI America™
CAS: 15484-80-7 Molecular Formula: C13H16O2 Molecular Weight (g/mol): 204.269 MDL Number: MFCD00080692 InChI Key: ZLHVSEPPILCZHH-UHFFFAOYSA-N Synonym: 4-tert-Butylbenzoic Acid Vinyl Ester PubChem CID: 84940 IUPAC Name: ethenyl 4-tert-butylbenzoate SMILES: CC(C)(C)C1=CC=C(C=C1)C(=O)OC=C