accessibility menu, dialog, popup

UserName

NaNaOHONOOOO

CAS RN 15467-20-6

IUPAC Name: disodium 2-[(carboxylatomethyl)(carboxymethyl)amino]acetate
Synonyms: disodium 2-[(carboxylatomethyl)(carboxymethyl)amino]acetate
Molecular Weight (g/mol): 235.1
Molecular Formula: C6H7NNa2O6
InChi Key: RHPXYIKALIRNFA-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC(=O)CN(CC([O-])=O)CC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results