accessibility menu, dialog, popup

UserName

NHNOFFF

CAS RN 1546-78-7

IUPAC Name: 6-(trifluoromethyl)-1,4-dihydropyrimidin-4-one
Synonyms: 6-(trifluoromethyl)-1H-pyrimidin-4-one
Molecular Weight (g/mol): 164.09
Molecular Formula: C5H3F3N2O
InChi Key: LVOYSBZJJWPUBD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)N=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results