accessibility menu, dialog, popup

UserName

SOOH2N

CAS RN 154350-28-4

IUPAC Name: cyclopropanesulfonamide
Synonyms: cyclopropanesulfonamide
Molecular Weight (g/mol): 121.15
Molecular Formula: C3H7NO2S
InChi Key: WMSPXQIQBQAWLL-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results