accessibility menu, dialog, popup

UserName

CAS RN 1540-36-9

IUPAC Name: 3-butylpentane-2,4-dione
Synonyms: 3-butylpentane-2,4-dione
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: MBXOOYPCIDHXGH-UHFFFAOYSA-N
SMILES: CCCCC(C(C)=O)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)