CAS RN 154-93-8
CAS RN 154-93-8
IUPAC Name:
1,3-bis(2-chloroethyl)-3-nitrosourea
Synonyms:
carmustine bcnu (bis chloroethylnitrosourea) carmustinum FIVB bis(chloroethyl) nitrosourea bcnu - carmustine C5H9cl2N3O2 carmustine preparation camustine bicnu (TN)
Molecular Weight (g/mol):
214.05
Molecular Formula:
C5H9Cl2N3O2
InChi Key:
DLGOEMSEDOSKAD-UHFFFAOYSA-N
SMILES:
ClCCNC(=O)N(CCCl)N=O
1
–
1
of
1
results
1
Carmustine 98.0+%, TCI America™
CAS: 154-93-8 Molecular Formula: C5H9Cl2N3O2 Molecular Weight (g/mol): 214.05 MDL Number: MFCD00057706 InChI Key: DLGOEMSEDOSKAD-UHFFFAOYSA-N Synonym: 1,3-Bis(2-chloroethyl)-1-nitrosourea PubChem CID: 2578 ChEBI: CHEBI:3423 IUPAC Name: 1,3-bis(2-chloroethyl)-3-nitrosourea SMILES: ClCCNC(=O)N(CCCl)N=O