accessibility menu, dialog, popup

UserName

ClClPtH3CSH3CCH3SCH3

CAS RN 15337-84-5

IUPAC Name: bis((ethylsulfanyl)ethane); dichloroplatinumbis(ylium)
Synonyms: dichloroplatinumbis(ylium); bis(ethyl sulfide)
Molecular Weight (g/mol): 446.35
Molecular Formula: C8H20Cl2PtS2
InChi Key: RFSLRYGKHBDFGC-UHFFFAOYSA-L
SMILES: Cl[Pt++]Cl.CCSCC.CCSCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results