accessibility menu, dialog, popup

UserName

CAS RN 15295-69-9

IUPAC Name: hept-6-ynenitrile
Synonyms: hept-6-ynenitrile
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: BIGRRVLEKCQFHS-UHFFFAOYSA-N
SMILES: C#CCCCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results