accessibility menu, dialog, popup

UserName

CAS RN 1528-41-2

H3COON

CAS RN 1528-41-2

IUPAC Name: ethyl 2-(4-cyanophenyl)acetate
Synonyms: ethyl 2-(4-cyanophenyl)acetate
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: DFEWKWBIPMKGFG-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results