accessibility menu, dialog, popup

UserName

CH3S

CAS RN 1518-75-8

IUPAC Name: 3-propylthiophene
Synonyms: 3-propylthiophene
Molecular Weight (g/mol): 126.22
Molecular Formula: C7H10S
InChi Key: QZNFRMXKQCIPQY-UHFFFAOYSA-N
SMILES: CCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results