accessibility menu, dialog, popup

UserName

ONOONHNH2OFFFHO

CAS RN 151038-94-7

IUPAC Name: 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanehydrazide; trifluoroacetic acid
Synonyms: 6-(2,5-dioxopyrrol-1-yl)hexanehydrazide; trifluoroacetic acid
Molecular Weight (g/mol): 339.27
Molecular Formula: C12H16F3N3O5
InChi Key: DRURMGRBAVYWCL-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)F.NNC(=O)CCCCCN1C(=O)C=CC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results