accessibility menu, dialog, popup

UserName

CAS RN 150993-62-7

H3CNCH3SOON(R)ClONON

CAS RN 150993-62-7

IUPAC Name: (2R)-1-[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl]pyrrolidine-2-carbonyl chloride
Synonyms: (2R)-1-[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl]pyrrolidine-2-carbonyl chloride
Molecular Weight (g/mol): 358.8
Molecular Formula: C13H15ClN4O4S
InChi Key: ZPBKQTOHYMFDLD-SECBINFHSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(N2CCC[C@@H]2C(Cl)=O)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results