accessibility menu, dialog, popup

UserName

CAS RN 15029-32-0

IUPAC Name: 3-(morpholin-4-yl)-3-oxopropanenitrile
Synonyms: morpholine, 4-(cyanoacetyl)-
Molecular Weight (g/mol): 154.17
Molecular Formula: C7H10N2O2
InChi Key: AUZPMUJGZZSMCP-UHFFFAOYSA-N
SMILES: O=C(CC#N)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results