accessibility menu, dialog, popup

UserName

CAS RN 14984-34-0

IUPAC Name: sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate
Synonyms: sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate
Molecular Weight (g/mol): 216.12
Molecular Formula: C6H9NaO7
InChi Key: WNFHGZLVUQBPMA-JSCKKFHOSA-M
SMILES: [Na+].O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)