accessibility menu, dialog, popup

UserName

CAS RN 149809-43-8

H3CSOOOONNNFF

CAS RN 149809-43-8

IUPAC Name: [5-(2,4-difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms: [5-(2,4-difluorophenyl)-5-(1,2,4-triazol-1-ylmethyl)oxolan-3-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 449.47
Molecular Formula: C21H21F2N3O4S
InChi Key: DFWVLCJRFGIRAK-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1COC(CN2C=NC=N2)(C1)C1=C(F)C=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results