accessibility menu, dialog, popup

UserName

CAS RN 1498-88-0

H3CNCH3H3CONOO

CAS RN 1498-88-0

IUPAC Name: 1',3',3'-trimethyl-6-nitro-1',3'-dihydrospiro[chromene-2,2'-indole]
Synonyms: 1',3',3'-trimethyl-6-nitrospiro[chromene-2,2'-indole]
Molecular Weight (g/mol): 322.36
Molecular Formula: C19H18N2O3
InChi Key: PSXPTGAEJZYNFI-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C(C=C2C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results