accessibility menu, dialog, popup

UserName

CAS RN 1496536-69-6

CH3OOOOOCH3OOCH3OOH3CH3C

CAS RN 1496536-69-6

IUPAC Name: [6-(4-methoxyphenoxy)-2,2-dimethyl-7-(4-methylbenzoyloxy)-hexahydro-[1,3]dioxolo[4,5-c]pyran-4-yl]methyl 4-methylbenzoate
Synonyms: [6-(4-methoxyphenoxy)-2,2-dimethyl-7-(4-methylbenzoyloxy)-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-4-yl]methyl 4-methylbenzoate
Molecular Weight (g/mol): 562.62
Molecular Formula: C32H34O9
InChi Key: BIJBLFRQSLTKJV-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC(=O)C3=CC=C(C)C=C3)C3OC(C)(C)OC3C2OC(=O)C2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results