accessibility menu, dialog, popup

UserName

CAS RN 14852-14-3

NHHNOOHOHOOH

CAS RN 14852-14-3

IUPAC Name: 2-hydroxybutanedioic acid; piperazine
Synonyms: malic acid; piperazine
Molecular Weight (g/mol): 220.23
Molecular Formula: C8H16N2O5
InChi Key: MGQUENZFJVXCFB-UHFFFAOYNA-N
SMILES: C1CNCCN1.OC(CC(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results