accessibility menu, dialog, popup

UserName

H3CCH3SiCH3CH3H3CH3CNBr

CAS RN 148249-36-9

IUPAC Name: 3-bromo-1-[tris(propan-2-yl)silyl]-1H-indole
Synonyms: 3-bromo-1-(triisopropylsilyl)indole
Molecular Weight (g/mol): 352.39
Molecular Formula: C17H26BrNSi
InChi Key: VELAHBNAGHGPNP-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=C(Br)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results