CAS RN 148-82-3
IUPAC Name:
2-amino-3-{4-[bis(2-chloroethyl)amino]phenyl}propanoic acid
Synonyms:
alkeran sarcolysine levofolan L-sarkolysin sarkolysin levofalan sarcolysine hydrochloride levopholan mustard, phenylalanine ccris 374
Molecular Weight (g/mol):
305.2
Molecular Formula:
C13H18Cl2N2O2
InChi Key:
SGDBTWWWUNNDEQ-UHFFFAOYNA-N
SMILES:
NC(CC1=CC=C(C=C1)N(CCCl)CCCl)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Melphalan, 94%
Melphalan, 94%
CAS: 148-82-3 Molecular Formula: C13H18Cl2N2O2 Molecular Weight (g/mol): 305.2 InChI Key: SGDBTWWWUNNDEQ-LBPRGKRZSA-N Synonym: melphalan,alkeran,melfalan,phenylalanine mustard,l-sarcolysine,l-pam,l-sarcolysin,l-phenylalanine mustard,levofalan,p-l-sarcolysin PubChem CID: 460612 ChEBI: CHEBI:28876 IUPAC Name: (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid SMILES: C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl