accessibility menu, dialog, popup

UserName

HNNHClHCl

CAS RN 147740-02-1

IUPAC Name: 5H,6H,7H-pyrrolo[3,4-b]pyridine dihydrochloride
Synonyms: 5H,6H,7H-pyrrolo[3,4-b]pyridine dihydrochloride
Molecular Weight (g/mol): 193.07
Molecular Formula: C7H10Cl2N2
InChi Key: FZBCVKVGLQRBHY-UHFFFAOYSA-N
SMILES: Cl.Cl.C1NCC2=C1C=CC=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results