accessibility menu, dialog, popup

UserName

CH3OOHO(Z)

CAS RN 14737-91-8

IUPAC Name: (2Z)-3-(2-methoxyphenyl)prop-2-enoic acid
Synonyms: (2Z)-3-(2-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: FEGVSPGUHMGGBO-SREVYHEPSA-N
SMILES: COC1=CC=CC=C1\C=C/C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results