accessibility menu, dialog, popup

UserName

CAS RN 14733-73-4

BrOOHN

CAS RN 14733-73-4

IUPAC Name: 5-bromo-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 2-benzoxazolinone, 5-bromo-
Molecular Weight (g/mol): 214.02
Molecular Formula: C7H4BrNO2
InChi Key: DMHTZWJRUUOALC-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OC(=O)NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results